Pyridinium p-toluenesulfonate, 98%
Pyridinium p-toluenesulfonate, 98%
Pyridinium p-toluenesulfonate, 98%
Thermo Scientific Chemicals

Pyridinium p-toluenesulfonate, 98%

CAS: 24057-28-1 | C12H13NO3S | 251.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 213740250
Price (USD)
75.70
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.70
Each
Chemical Identifiers
CAS24057-28-1
IUPAC Name4-methylbenzene-1-sulfonic acid; pyridine
Molecular FormulaC12H13NO3S
InChI KeyZDYVRSLAEXCVBX-UHFFFAOYSA-N
SMILESC1=CC=NC=C1.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point112°C to 119°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only