3-Aminoquinoline, 99%
3-Aminoquinoline, 99%
3-Aminoquinoline, 99%
Thermo Scientific Chemicals

3-Aminoquinoline, 99%

CAS: 580-17-6 | C9H8N2 | 144.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 213780050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS580-17-6
IUPAC Namequinolin-3-amine
Molecular FormulaC9H8N2
InChI KeySVNCRRZKBNSMIV-UHFFFAOYSA-N
SMILESNC1=CN=C2C=CC=CC2=C1
さらに表示
Melting point92°C to 96°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to beige to orange-brown
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only