ART™ Barrier Low-Retention Pipette Tips
Thermo Scientific™
ART™ Barrier Low-Retention Pipette Tips
Eliminate sample retention with these specially designed pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelTip StyleFit for PurposeUnit Size
2139-0510 μLART 10Low Retention, Micro PointSterile and PCR ReadyCase of 4800
2139-05PK
also known as 2139-05
10 μLART 10Low Retention, Micro PointSterile and PCR ReadyPack of 960
2140-0510 μLART 10 REACHExtended Length, Low Retention, Micro PointSterile and PCR ReadyCase of 4800
2140-05PK10 μLART 10 REACHExtended Length, Low Retention, Micro PointSterile and PCR ReadyPack of 960
2149P-0520 μLART 20PLow Retention, Micro PointSterile and PCR ReadyCase of 4800
2149P-05PK
also known as 2149P-05
20 μLART 20PLow Retention, Micro PointSterile and PCR ReadyPack of 960
2069-05200 μLART 200Low Retention, Micro PointSterile and PCR ReadyCase of 4800
2069-05PK
also known as 2069-05
200 μLART 200Low Retention, Micro PointSterile and PCR ReadyPack of 960
2279-051000 μLART 1000Ergonomic, Low RetentionSterile and PCR ReadyCase of 3200
2279-05PK
also known as 2279-05
1000 μLART 1000Ergonomic, Low RetentionSterile and PCR ReadyPack of 800
Catalog number 2139-05
Price (USD)
700.00
Online Exclusive
1,000.00
Save 300.00 (30%)
Case of 4800
Volume (Metric):
10 μL
Model:
ART 10
Tip Style:
Low Retention, Micro Point
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 4800
Price (USD)
700.00
Online Exclusive
1,000.00
Save 300.00 (30%)
Case of 4800
Get Thermo Scientific™ ART™ Barrier Low-Retention Pipette Tips, which are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • Certified RNase, DNase, ATP, and Endotoxin Free
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 10, Filtered, Sterile, Rack
ModelART 10
PackagingRack, 10 x 96
AutoclavableNot Autoclavable
ColorClear
FilteredFiltered
Fit for PurposeSterile and PCR Ready
FormatRack
Product LineART
SterilitySterile
Tip StyleLow Retention, Micro Point
Volume (Metric)10 μL
Unit SizeCase of 4800