Glycerine, 99.0-101.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Glycerine, 99.0-101.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, EP, BP, and FCC specifications.
Have Questions?
KatalognummerMenge
2140-0R550 lb.
Katalognummer 2140-0R
Preis (EUR)
-
Menge:
550 lb.
This product is GMP Manufactured and meets USP, EP, BP, and FCC specifications.
This product is GMP Manufactured and meets USP, EP, BP, and FCC specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
FarbeFarblos
Schmelzpunkt18.6°C
VerpackungPolyethylene drum
Siedepunkt290°C (1013 hPa)
GüteMulti-Kompendium/USP
Molekulargewicht (g/mol)92.09
Reinheit (%)99 to 101%
Physikalische FormLiquid
Menge550 lb.
Unit Size550 lb.