Glycerine (vegetable based), 99.0-101.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Glycerine (vegetable based), 99.0-101.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2143-01500 mL
2143-0719 L
2143-09200 L
2143-034 L
Catalog number 2143-01
Price (MXN)
-
Quantity:
500 mL
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorColorless
Melting Point18.6°C
PackagingAmber glass bottle, narrow mouth
Boiling Point290°C (1013 hPa)
GradeMulti-Compendial/USP
Molecular Weight (g/mol)92.09
Percent Purity99 to 101%
Physical FormLiquid
Quantity500 mL
Unit SizeEach