Product Image
Gibco™
Difco™ PPLO Broth, porcine H
Difco PPLO Broth, porcine H, is used for isolating and cultivating Mycoplasma when supplemented with nutritive enrichments. Meat digests, peptones,Weitere Informationen
Have Questions?
KatalognummerMenge
21492810 kg
Katalognummer 214928
Preis (EUR)
0,00
Each
Menge:
10 kg
Preis (EUR)
0,00
Each
Difco PPLO Broth, porcine H, is used for isolating and cultivating Mycoplasma when supplemented with nutritive enrichments. Meat digests, peptones, beef extract, and yeast extract provide the nitrogen, vitamins, amino acids and carbon in these media.
Specifications
Menge10 kg
FormDehydriertes Pulver
SpeziesPorcine
Unit SizeEach
Inhalt und Lagerung
Lagerbedingungen: 2 bis 25 °C. Kühl und trocken in einem gut versiegelten Behälter lagern.