ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
ART™ Barrier Pipette Tips in Lift-off Lid Rack
Thermo Scientific™

ART™ Barrier Pipette Tips in Lift-off Lid Rack

Perform applications accurately and consistently with Barrier Pipette Tips in Lift-off Lid Racks, which seal when exposed to potential contaminants.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Model
2149P20 μLART 20
2070300 μLART 300
2065E100 μLART 100E
2069200 μLART 200
2065100 μLART 100
2079E1000 μLART 1000E
2139F0.5 to 10 μLART 10F
2149E10 μLART 10
Catalog number 2149P
Price (MXN)
-
Volume (Metric):
20 μL
Model:
ART 20
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Pipette Tips in Lift-off Lid Rack, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier pipette tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 20, Filtered, Sterile, Lift-off Lid Rack
Length (English)2 in.
Length (Metric)5.07 cm
ModelART 20
Volume (Metric) Dispensing20μL
AutoclavableNot Autoclavable
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleMicro Point
Volume (Metric)20 μL
Unit SizeCase of 4800