ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Thermo Scientific™

ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks

The pipette tip and rack mail-back program provides a simple and effective solution for recycling PP#5 single use plastics. The TipCycle Kit included with your case of pipette tips to collect used pipette tips and empty racks. Complete waiver, attach pre-paid shipping label and return for recycling.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoVolumen (métrico)Estilo de puntaModelo
2149PRC-05-HR20 μLRetención bajaART 20 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2065ERC-HR100 μLEstándarART 100 μl, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2069RC-05-HR200 μLRetención bajaART 200 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2139RC-05-HR10 μLRetención bajaART 10 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2139RC-HR10 μLEstándarART 10 μl, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2179RC-HR1000 μLEstándarART 1000 μl, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2069RC-HR200 μLEstándarART 200 μl, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2149PRC-HR20 μLEstándarART 20 μl, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
2065ERC-05-HR100 μLRetención bajaART 100 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
Número de catálogo 2149PRC-05-HR
Precio (USD)
-
Volumen (métrico):
20 μL
Estilo de punta:
Retención baja
Modelo:
ART 20 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle

Unlike standard length barrier tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks self-seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in MicroPoint™, low retention and offer a universal fit design.

Features:

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
  • Stackable hinged racks save space in your lab
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Lid rim prevents racks from sliding around when stacked and carted around a lab
Especificaciones
Certificaciones/conformidadISO 9001:2015
Declaraciones de limpiezaSin ADNasa, ARNasa, ADN, ATP, inhibidor de PCR ni endotoxinas
DesechablesSingle-use
Para utilizar con (aplicación)Liquid Handling, Molecular Biology, PCR
Incluye1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Longitud (imperial)2 in
Longitud (métrico)5.07 cm
ModeloART 20 μl, baja retención, gradilla estéril de barrera con bisagras, kit de reciclaje TipCycle
EnvaseHinged Rack
Tipo de productoTipCycle, Pipette Tip and Rack Recycling Kit
Cantidad4800/Cs.
Dispensación por volumen (métrico)20 μL
Esterilizables en autoclaveNo Autoclavable
ColorNatural
filtradas porCon filtro
Fit for PurposeSterile and PCR Ready
FormatoHinged Rack
Línea de productosART
EsterilidadEstéril
Estilo de puntaRetención baja
Volumen (métrico)20 μL
Unit SizeCase of 4800