Methylenediphosphonic acid, 98%, Thermo Scientific Chemicals
Methylenediphosphonic acid, 98%, Thermo Scientific Chemicals
Methylenediphosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methylenediphosphonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 215070050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1984-15-2
IUPAC Name(phosphonomethyl)phosphonic acid
Molecular FormulaCH6O6P2
InChI KeyMBKDYNNUVRNNRF-UHFFFAOYSA-N
SMILESOP(O)(=O)CP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Color)White
Melting point197°C to 204°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only