PIPES, disodium salt, 99%
PIPES, disodium salt, 99%
PIPES, disodium salt, 99%
Thermo Scientific Chemicals

PIPES, disodium salt, 99%

CAS: 76836-02-7 | C8H16N2Na2O6S2 | 346.324 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 215090250
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.40
Save 9.75
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.40
Save 9.75
Each
Chemical Identifiers
CAS76836-02-7
IUPAC Namedisodium 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H16N2Na2O6S2
InChI KeyGMHSTJRPSVFLMT-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water=<5 %
UV(0.1 M in water)
UVat 280 nm A: =<0.050
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only