Potassium indigotrisulfonate
Potassium indigotrisulfonate
Potassium indigotrisulfonate
Thermo Scientific Chemicals

Potassium indigotrisulfonate

CAS: 67627-18-3 | C16H7K3N2O11S3 | 616.71 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 215170250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67627-18-3
IUPAC Nametripotassium (Z)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
Molecular FormulaC16H7K3N2O11S3
InChI KeyXOSMXDUITYWYGR-JRYLAINFSA-K
SMILES[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2NC(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O
さらに表示
Appearance (Color)Dark blue
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Molar extinction coefficient0.01 g/l aq.soln. (1 cm cell)
Molar extinction coefficient>= 15000 (597-603 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only