Thermo Scientific™

DFDNB (1,5-difluoro-2,4-dinitrobenzene)

Catalog number: 21525
Thermo Scientific™

DFDNB (1,5-difluoro-2,4-dinitrobenzene)

Catalog number: 21525
Catalog Number
21525
Unit Size
50 mg
Price (USD)
78.50
Your Price:
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
2152550 mg
78.50
Your Price:
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
Thermo Scientific Pierce DFDNB is 1,5-difluoro-2,4-dinitrobenzene, an aryl halide compound with two reactive fluorine atoms that can couple to amine-containing molecules, yielding stable arylamine bonds.

Features of DFDNB:

Reactive groups: aryl halide (both ends)
Reactive towards: amino groups (primary amines)
• Short distance crosslinks

Product References
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homobifunctional
Reactive Moiety
Active Ester, TFP Ester
Spacer
Short
Form
Powder
Quantity
50 mg
Type
Crosslinker
Solubility
DMF, DMSO

Contents & Storage

Store at 4°C.

Figures