1-Hexanethiol, 96%
1-Hexanethiol, 96%
1-Hexanethiol, 96%
Thermo Scientific Chemicals

1-Hexanethiol, 96%

CAS: 111-31-9 | C6H14S | 118.24 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
產品號碼 215275000
價格 (TWD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS111-31-9
IUPAC Namehexane-1-thiol
Molecular FormulaC6H14S
InChI KeyPMBXCGGQNSVESQ-UHFFFAOYSA-N
SMILESCCCCCCS
檢視更多
規格Specification Sheet規格表
Refractive index1.4480 to 1.4520 (20°C, 589 nm)
GC>=95 %
Density(20°C) 0.836 to 0.844 g/ml
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only