Citric acid, monosodium salt, 99%, pure, anhydrous
Citric acid, monosodium salt, 99%, pure, anhydrous
Citric acid, monosodium salt, 99%, pure, anhydrous
Thermo Scientific Chemicals

Citric acid, monosodium salt, 99%, pure, anhydrous

CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 215330025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS18996-35-5
IUPAC Namesodium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Molecular FormulaC6H7NaO7
InChI KeyHWPKGOGLCKPRLZ-UHFFFAOYSA-M
SMILES[Na+].OC(=O)CC(O)(CC(O)=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only