Soy 100
Gibco™

Soy 100

Soy 100 is a highly digested soy protein peptone that is high in nucleosides and free amino acid content. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
215383100 g
670138500 g
67013710 kg
Catalog number 215383
Price (MXN)
-
Quantity:
100 g

Soy 100 is a highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates, which is recommended for the cultivation of a wide variety of microbial organisms as well as varied mammalian cell culture applications.

Soy 100 can be blended with other animal origin-free peptones, such as wheat or yeast, when looking for replacements for animal origin peptones.

Soy 100 is also part of the Gibco Peptone Preview Pak, which features our new wheat, cotton, soy, and yeast peptones. These peptones provide unique nutritional profiles compared to other cell culture supplements. The Preview Pak includes five 100 gram samples*.

*The Gibco Peptone Preview Pak is non-GMP. Larger GMP packaging sizes are available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Cell Culture and Microbial Fermentation, Tissue Culture Application
FormatBottle, Pail
Product LineGibco
Quantity100 g
RemarksThere are no animal sourced ingredients or animal derived reagents used in the manufacture of this product.
Shelf Life2 Years
SourceAnimal origin-free
FormPowder
Product TypeDehydrated Culture Media
Regulatory StatusResearch Use or Further Manufacturing
pH6.2 to 7.6
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.