Product Image
Gibco™

CMRL Medium (10X), no glutamine

CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L' cells under serum-free conditions.深入閱讀
Have Questions?
產品號碼Quantity
21540026500 mL
產品號碼 21540026
價格 (TWD)
19,530.00
線上優惠
Ends: 30-Sep-2026
27,900.00
您節省 8,370.00 (30%)
Each
Quantity:
500 mL
Customize this product
價格 (TWD)
19,530.00
線上優惠
Ends: 30-Sep-2026
27,900.00
您節省 8,370.00 (30%)
Each

CMRL Medium was originally developed by Connaught Medical Research Laboratories for the growth of Earle's 'L' cells under serum-free conditions. Gibco™ CMRL Medium is also especially useful for cloning monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.

This CMRL Medium is modified as follows:
WithWithout
• Phenol Red• L-glutamine
 • HEPES
 • Sodium Bicarbonate


The complete formulation is available.

Gibco™ CMRL Medium is a modified version of Medium 199. CMRL medium is particularly rich in nucleosides and vitamins.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

CMRL Medium contains no proteins or growth factors. Therefore, CMRL Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). CMRL Medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
規格
Cell TypeMammalian Cells
Concentration10 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeCMRL Medium
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesLow Glucose
Without AdditivesNo Sodium Pyruvate, No Sodium Bicarbonate
Unit SizeEach
內容物與存放
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

常見問答集 (常見問題)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.