1-Bromohexadecane, 97%
1-Bromohexadecane, 97%
1-Bromohexadecane, 97%
Thermo Scientific Chemicals

1-Bromohexadecane, 97%

CAS: 112-82-3 | C16H33Br | 305.34 g/mol
数量:
250 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 215430025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS112-82-3
Appearance (Form)Liquid or solid
Appearance (Color)Clear colorless to yellow to amber to brown
Infrared spectrumConforms
Refractive index1.4600 to 1.4640 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only