ART™ Gel Loading Pipette Tips
Thermo Scientific™
ART™ Gel Loading Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Fit for PurposeUnit Size
2155100 μLSterile and PCR ReadyCase of 3840
2155PK
also known as 2155
100 μLSterile and PCR ReadyPack of 960
2155P20 μLSterile and PCR ReadyCase of 3840
2155PPK20 μLSterile and PCR ReadyPack of 960
Catalog number 2155
Price (USD)
662.20
Online exclusive
880.00
Save 217.80
Case of 3840
Volume (Metric):
100 μL
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 3840
Price (USD)
662.20
Online exclusive
880.00
Save 217.80
Case of 3840
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Gel Loading Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications.
Features:
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Gel loading for loading acrylamide or agarose gels
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART Gel 100, Filtered, Sterile, Lift-off Lid Rack
Length (English)2.79 in.
Length (Metric)7.09cm
ModelART Gel 100
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Gilson Pipetman, Thermo Scientific Finnpipette Focus
FormatLift-off Lid Rack
Product LineART
SterilitySterile
Tip StyleGel Loading
Volume (Metric)100 μL
Unit SizeCase of 3840