2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals

2-(Dimethylamino)ethyl methacrylate, 99%, stabilized

CAS: 2867-47-2 | C8H15NO2 | 157.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 215840010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2867-47-2
IUPAC Name2-(dimethylamino)ethyl 2-methylprop-2-enoate
Molecular FormulaC8H15NO2
InChI KeyJKNCOURZONDCGV-UHFFFAOYSA-N
SMILESCN(C)CCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
Stabilizer750 to 2000 ppm MEHQ
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only