ART Reach Barrier Tips, Low Retention
Molecular Bio Products Inc.
ART Reach Barrier Tips, Low Retention
Precise pipetting with reduced sample loss.
Have Questions?
Catalog NumberVolume (Metric)
2160P-05-HR200 μL
Catalog number 2160P-05-HR
Price (EUR)
812,00
Case of 3840
Volume (Metric):
200 μL
Price (EUR)
812,00
Case of 3840
Precise pipetting with reduced sample loss.
  • Low retention
  • Barrier filter
  • Extended reach
  • Sterile
  • Accurate
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART Barrier, Filtered, Sterile, Hinged Rack
Length (English)3.6 in.
Length (Metric)9.09 cm
ModelART Barrier
Product TypePipette Tips
Quantity768 Tips
AutoclavableNot Autoclavable
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleExtended Length, Low Retention
Volume (Metric)200 μL
Unit SizeCase of 3840