1,4-Dichlorobutane, 99%
1,4-Dichlorobutane, 99%
1,4-Dichlorobutane, 99%
Thermo Scientific Chemicals

1,4-Dichlorobutane, 99%

CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol
数量:
100 mL
500 mL
1 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 216435000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-56-5
IUPAC Name1,4-dichlorobutane
Molecular FormulaC4H8Cl2
InChI KeyKJDRSWPQXHESDQ-UHFFFAOYSA-N
SMILESClCCCCCl
さらに表示
Water=<0.05 %
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only