Octamethylcyclotetrasiloxane, 98%
Octamethylcyclotetrasiloxane, 98%
Octamethylcyclotetrasiloxane, 98%
Thermo Scientific Chemicals

Octamethylcyclotetrasiloxane, 98%

CAS: 556-67-2 | C8H24O4Si4 | 296.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 216471000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS556-67-2
IUPAC Nameoctamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular FormulaC8H24O4Si4
InChI KeyHMMGMWAXVFQUOA-UHFFFAOYSA-N
SMILESC[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.3950 to 1.3970 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only