Product Image
Gibco™

Glasgow's MEM

Glasgow's Minimum Essential Medium (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's medium.Read more
Have Questions?
Catalog NumberQuantity
21710082
also known as 21710-082
10 x 500 mL
Catalog number 21710082
also known as 21710-082
Price (MXN)
-
Quantity:
10 x 500 mL
Customize this product

Glasgow's Minimum Essential Medium (GMEM) was originally developed by Ian McPherson and Michael Stoker as a modification of Eagle's medium. It was utilized to study the genetic factors which affected cell competence. Glasgow's MEM was developed for use with kidney cells lines, such as BHK-21.

This MEM is modified as follows:

With: L-glutamine, Phenol Red

Without: Tryptose phosphate broth, HEPES

The complete formulation is available.

Glasgow's MEM is unique from other media as it contains twice the concentration of amino acids and vitamins than the original Basal Medium Eagle, and is used without serum. Glasgow's MEM was originally formulated with 10% tryptose phosphate broth.

Glasgow's MEM contains no proteins, lipids, or growth factors. Therefore, Glasgow's MEM requires supplementation, commonly with 10% tryptose phosphate broth. Glasgow's MEM uses a sodium bicarbonate buffer system (2.75 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Specifications
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMinimum Essential Medium (MEM)
Quantity10 x 500 mL
Shelf Life12 Months
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Phenol Red
Without AdditivesNo HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture