This product is the replacement for Catalog No. 11720-067 and 11720-034. The formulations are identical, made with slightly different yeastolates derived from the same species. For more information, please contact our Technical Service team at cellculturesupport@lifetech.com.
Schneider's <i>Drosophila</i> Medium
Gibco™

Schneider's Drosophila Medium

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's DrosophilaRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21720024500 mL
2172000110 x 500 mL
Catalog number 21720024
Price (USD)
95.65
Online exclusive
102.00
Save 6.35
Each
Quantity:
500 mL
Customize this product
Price (USD)
95.65
Online exclusive
102.00
Save 6.35
Each
Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells. We offer a variety of Gibco™ Media options for a range of insect cell culture applications.

Formulation
This Schneider's Drosophila Medium is modified; it contains L-glutamine and sodium bicarbonate. The complete formulation is available. Schneider's Drosophila Medium contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelose.

cGMP manufacturing and quality system
Gibco™ Schneider’s Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Schneider's Drosophila Medium is commonly supplemented with 10% fetal bovine serum (FBS) before use. Schneider's Drosophila Medium requires ambient CO2 in order to maintain physiological pH.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineS2 (Schneider 2)
Cell TypeInsect Cell
Product LineGibco
Product TypeSchneider's Drosophila Medium
Quantity500 mL
Shipping ConditionRoom Temperature
FormLiquid
With AdditivesGlutamine
Without AdditivesNo HEPES, No Phenol Red, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Are D. Mel-2 cells the same as Schneider S2 cells?

D. Mel-2 (Drosophila melanogaster-2) cells are basically Schneider S2 cells that have been adapted to growth in serum-free media.