Aluminum chloride hexahydrate, 99%, pure
Aluminum chloride hexahydrate, 99%, pure
Aluminum chloride hexahydrate, 99%, pure
Thermo Scientific Chemicals

Aluminum chloride hexahydrate, 99%, pure

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 217470025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7784-13-6
IUPAC Namealuminium(3+) hexahydrate trichloride
Molecular FormulaAlCl3H12O6
InChI KeyJGDITNMASUZKPW-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Adhering powder or crystals
Titration Argentometric>=98.5 %
Water42 to 48 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only