Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis
Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis
Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis

CAS: 7784-24-9 | AlKO8S2·12H2O | 474.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 217480010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7784-24-9
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
pH3 to 3.5 (10 % at 20°C)
Chloride (Cl)=<50 ppm
Iron (Fe)=<50 ppm
Arsenic (As)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only