Calcium nitrate tetrahydrate, 99%, for analysis
Calcium nitrate tetrahydrate, 99%, for analysis
Calcium nitrate tetrahydrate, 99%, for analysis
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 99%, for analysis

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 217510010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13477-34-4
Molecular FormulaCaH8N2O10
InChI KeyMWGCGFYACDTFSB-UHFFFAOYSA-N
SMILESO.[Ca++].[O-][N+]([O-])=O
Molecular Weight (g/mol)236.15
SpecificationsSpecification SheetSpecification Sheet
pH4.5 to 7.0 (5 % at 25°C)
Appearance (Color)White
Appearance (Form)Crystals
Chloride (Cl)=<50 ppm
Titration Complexometric98.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only