(S)-(-)-sec-Phenethyl alcohol, 99%
(S)-(-)-sec-Phenethyl alcohol, 99%
(S)-(-)-sec-Phenethyl alcohol, 99%
Thermo Scientific Chemicals

(S)-(-)-sec-Phenethyl alcohol, 99%

CAS: 1445-91-6 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 217790050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1445-91-6
IUPAC Name(1S)-1-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWAPNOHKVXSQRPX-ZETCQYMHSA-N
SMILESC[C@H](O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-41° to -44° (20°C, 589 nm) (neat)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5260 to 1.5280 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only