ART™ Barrier Extended Length Reload System
Thermo Scientific™
ART™ Barrier Extended Length Reload System
Perform applications accurately and consistently without compromising protection with this barrier reload system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTip StyleFit for PurposeUnit Size
2179-RTExtended LengthSterile and PCR ReadyCase of 2800
2179-05-RTLow Retention, Extended LengthSterile and PCR ReadyCase of 2800
2179-05-RTPK
also known as 2179-05-RT
Low Retention, Extended LengthSterile and PCR ReadyPack of 560
2179-RTPK
also known as 2179-RT
Extended LengthSterile and PCR ReadyPack of 560
Catalog number 2179-RT
Price (USD)
402.50
Online exclusive
575.00
Save 172.50
Case of 2800
Tip Style:
Extended Length
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 2800
Price (USD)
402.50
Online exclusive
575.00
Save 172.50
Case of 2800
Perform applications accurately and consistently without compromising protection with the Thermo ScientificTM ARTTM Barrier Reload System, which provides an environmentally-responsible solution without compromising sterility. With offset wells that allow the tips to stack, this advanced system reduces waste and saves bench top and storage space. The ART portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in your laboratory.
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART Barrier, Filtered, Sterile, Rack
Length (English)3.5 in.
Length (Metric)8.89 cm
ModelART Barrier
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizeCase of 2800