(S)-(+)-sec-Butanol, 99%
(S)-(+)-sec-Butanol, 99%
(S)-(+)-sec-Butanol, 99%
Thermo Scientific Chemicals

(S)-(+)-sec-Butanol, 99%

CAS: 4221-99-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 218030050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4221-99-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+12° to +14° (20°C, 589 nm) (neat)
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only