Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)

CAS: 13494-98-9 | N3O96H2O | 383.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 218261000
Price (EUR)
159,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
159,00
Each
Chemical Identifiers
CAS13494-98-9
IUPAC Nameyttrium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Y
InChI KeyQBAZWXKSCUESGU-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals
Total trace metal impurities=<0.1 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only