Acetic acid, ammonium salt, 98+%, for analysis
Acetic acid, ammonium salt, 98+%, for analysis
Acetic acid, ammonium salt, 98+%, for analysis
Thermo Scientific Chemicals

Acetic acid, ammonium salt, 98+%, for analysis

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 218365000
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.00
Save 12.35 (15%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.00
Save 12.35 (15%)
Each
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=98.0 % (on dry substance)
Ash=<0.01 %
Iron (Fe)=<20 ppm
Water=<3.0 %
pH6.5 to 7.5 (5 % at 20°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only