Polymyxin B Sulfate
Gibco™

Polymyxin B Sulfate

Polymyxin B Sulfate is produced by Bacillus polymyxa. Polymyxin B Sulfate works by binding to the cellular membrane and interfering자세히 알아보기
Have Questions?
카탈로그 번호수량
2185002925 MU
카탈로그 번호 21850029
제품 가격(KRW)
743,000
온라인 행사
Ends: 15-Jul-2026
825,000
할인액 82,000 (10%)
Each
수량:
25 MU
제품 가격(KRW)
743,000
온라인 행사
Ends: 15-Jul-2026
825,000
할인액 82,000 (10%)
Each
Polymyxin B Sulfate is produced by Bacillus polymyxa. Polymyxin B Sulfate works by binding to the cellular membrane and interfering with its permeability. Gibco™ Polymyxin B Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria. The recommended working concentration is 100 units/ml. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.
사양
농도100 Units/mL
용도(애플리케이션)Bacterial Selection
수량25 MU
유통 기한24 Months
배송 조건Room Temperature
형태Powder
제품 유형Antibiotic
멸균Non-sterile
Unit SizeEach
구성 및 보관
Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

자주 묻는 질문(FAQ)

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

인용 및 참조 문헌 (2)

인용 및 참조 문헌
Abstract
The tumour suppressor CDKN2A/p16
Authors:Wouters K, Deleye Y, Hannou SA, Vanhoutte J, Maréchal X, Coisne A, Tagzirt M, Derudas B, Bouchaert E, Duhem C, Vallez E, Schalkwijk CG, Pattou F, Montaigne D, Staels B, Paumelle R
Journal:Diab Vasc Dis Res
PubMed ID:28868898
The genomic CDKN2A/B locus, encoding p16
Progranulin in the hematopoietic compartment protects mice from atherosclerosis.
Authors:Nguyen AD, Nguyen TA, Singh RK, Eberlé D, Zhang J, Abate JP, Robles A, Koliwad S, Huang EJ, Maxfield FR, Walther TC, Farese RV
Journal:Atherosclerosis
PubMed ID:30212683
Progranulin is a circulating protein that modulates inflammation and is found in atherosclerotic lesions. Here we determined whether inflammatory cell-derived progranulin impacts atherosclerosis development.