RPMI 1640 Medium
Actual product may vary
Gibco™

RPMI 1640 Medium

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21875158
also known as 21875-158
6 x 1000 mL
21875042
also known as 21875-042
1 x 100 mL
21875109
also known as 21875-109
20 x 100 mL
21875034
also known as 21875-034
1 x 500 mL
21875091
also known as 21875-091
10 x 500 mL
21875059
also known as 21875-059
1 x 1000 mL
Catalog number 21875158
also known as 21875-158
Price (EUR)
246,00
Each
Quantity:
6 x 1000 mL
Customize this product
Price (EUR)
246,00
Each

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:

  • With
    • Glutamine
    • Phenol Red
  • Without
    • HEPES

The complete formulation is available.

Using RPMI

RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell TypeMammalian Cells
Concentration1X
Culture EnvironmentCO2
For Use With (Application)Mammalian Cell Culture
FormatBottles
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 - 310 mOsm/kg
Product LineGibco
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity6 x 1000 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
ClassificationAnimal Origin-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red, Sodium Bicarbonate
Without AdditivesNo HEPES, No Sodium Pyruvate
pH7.0 to 7.4
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.