3-Methyl-2-butanone, 98%
3-Methyl-2-butanone, 98%
3-Methyl-2-butanone, 98%
Thermo Scientific Chemicals

3-Methyl-2-butanone, 98%

CAS: 563-80-4 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Plastic drum
Catalog number 218770051
Price (USD)
629.00
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
629.00
Each
Chemical Identifiers
CAS563-80-4
IUPAC Name3-methylbutan-2-one
Molecular FormulaC5H10O
InChI KeySYBYTAAJFKOIEJ-UHFFFAOYSA-N
SMILESCC(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3855 to 1.3905 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only