Methyl isocyanoacetate, 95%, Tech.
Methyl isocyanoacetate, 95%, Tech.
Methyl isocyanoacetate, 95%, Tech.
Thermo Scientific Chemicals

Methyl isocyanoacetate, 95%, Tech.

CAS: 39687-95-1 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 219000050
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Chemical Identifiers
CAS39687-95-1
IUPAC Namemethyl 2-(methylideneamino)acetate
Molecular FormulaC4H7NO2
InChI KeyZRGVTTCCASYOFN-UHFFFAOYSA-N
SMILESCOC(=O)CN=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4161 to 1.4191 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only