Aluminum hydroxide, extra pure, powder
Aluminum hydroxide, extra pure, powder
Aluminum hydroxide, extra pure, powder
Thermo Scientific Chemicals

Aluminum hydroxide, extra pure, powder

CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 219130020
Price (USD)
-
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS21645-51-2
IUPAC Namealuminium(3+) trihydroxide
Molecular FormulaAlH3O3
InChI KeyWNROFYMDJYEPJX-UHFFFAOYSA-K
SMILES[OH-].[OH-].[OH-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
Assay>=63.5 % (Al2O3)
Loss on ignition=<36.5 %
Soluble matter=<0.2 % (in water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only