Calcium carbonate, 99%, for analysis, precipitated
Calcium carbonate, 99%, for analysis, precipitated
Calcium carbonate, 99%, for analysis, precipitated
Thermo Scientific Chemicals

Calcium carbonate, 99%, for analysis, precipitated

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 219160010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White
Titration Complexometric>=98.5 % (on dried substance)
Sulfate (SO4)=<0.2 %
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only