Sodium sulfite, 98.5%, for analysis, anhydrous
Sodium sulfite, 98.5%, for analysis, anhydrous
Sodium sulfite, 98.5%, for analysis, anhydrous
Thermo Scientific Chemicals

Sodium sulfite, 98.5%, for analysis, anhydrous

CAS: 7757-83-7 | Na2O3S | 126.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 219272500
Price (USD)
38.90
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
38.90
Each
Chemical Identifiers
CAS7757-83-7
IUPAC Namedisodium heptahydrate sulfite
Molecular FormulaH14Na2O10S
InChI KeyWMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder or crystals
Titration Iodimetric>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only