Benzophenone, 99+%, pure
Benzophenone, 99+%, pure
Benzophenone, 99+%, pure
Thermo Scientific Chemicals

Benzophenone, 99+%, pure

CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 219680500
Price (USD)
19.65
Online exclusive
21.30
Save 1.65
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
19.65
Online exclusive
21.30
Save 1.65
Each
Chemical Identifiers
CAS119-61-9
IUPAC Namediphenylmethanone
Molecular FormulaC13H10O
InChI KeyRWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
GC>=99.0 %
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point47°C to 49°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only