Search
View additional product information for DMEM, high glucose, pyruvate, no glutamine - FAQs (21969035)
1 product FAQs found
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.