Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure, 500mL - FAQs
Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure, 500mL - FAQs
View additional product information for Nalgene™ Fluorinated Wide-Mouth HDPE Bottles with Closure - FAQs (2197-0008PK, 2197-0016, 2197-0004, 2197-0016PK, 2197-0032, 2197-0004PK, 2197-0008, 2197-0032PK)
This product does not have any FAQs available.