Thiourea, 99+%, for analysis
Thiourea, 99+%, for analysis
Thiourea, 99+%, for analysis
Thermo Scientific Chemicals

Thiourea, 99+%, for analysis

CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 220050010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Zinc (Zn)=<10 ppm
Sulfated ash=<0.1 %
Loss on drying=<0.5 %
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only