(±)-Propylene oxide, 99.5%, extra pure
(±)-Propylene oxide, 99.5%, extra pure
(±)-Propylene oxide, 99.5%, extra pure
Thermo Scientific Chemicals

(±)-Propylene oxide, 99.5%, extra pure

CAS: 75-56-9 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 220160050
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Chemical Identifiers
CAS75-56-9
IUPAC Name2-methyloxirane
Molecular FormulaC3H6O
InChI KeyGOOHAUXETOMSMM-UHFFFAOYNA-N
SMILESCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.4 %
Aldehyde (C3H6O)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only