Nalgene™ Large Narrow-Mouth LDPE Bottles
Nalgene™ Large Narrow-Mouth LDPE Bottles
Nalgene™ Large Narrow-Mouth LDPE Bottles
Nalgene™ Large Narrow-Mouth LDPE Bottles
Nalgene™ Large Narrow-Mouth LDPE Bottles
Thermo Scientific™

Nalgene™ Large Narrow-Mouth LDPE Bottles

Collect and store distilled water in bottles that offer flexibility, impact resistance and visibility of contents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2202-00104 L38-430 mmCase of 6
2202-00052 L38-430 mmCase of 6
2202-0005PK
also known as 2202-0005
2 L38-430 mmEach
2202-0010PK
also known as 2202-0010
4 L38-430 mmEach
2202-00208 L53BCase of 6
2202-0020PK
also known as 2202-0020
8 L53BEach
Catalog number 2202-0010
Price (EUR)
143,00
Case of 6
Capacity (Metric):
4 L
Closure Size:
38-430 mm
Unit Size:
Case of 6
Price (EUR)
143,00
Case of 6

Store chemicals and standards and collect and store distilled water. Thermo Scientific™ Nalgene™ Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low-density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contiminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Polypropylene screw caps, leakproof† when used with Nalgene bottles with the matching finish

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided

.

Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity1 gal., 4 L
Capacity (English)1 gal.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorWhite
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypeScrew Cap
DescriptionLarge Narrow-Mouth Bottle
Diameter (Metric)152 mm
Diameter (Metric) Inner Neck25 mm
Height (English)13.1 in.
Height (Metric)333 mm
Neck Finish38 mm
No. per Case6
No. per Pack1
Product TypeNarrow Mouth Bottle
ShapeRound
Through the Closure DispensingNo
Width (English)6 in.
Capacity (Metric)4 L
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialLDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 6