Biohazard Disposal Bags with or without Warning Label
Fisher Scientific™

Biohazard Disposal Bags with or without Warning Label

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
Have Questions?
카탈로그 번호치수(길이 x 폭)색상
22032926
FISHERMIS-22032926-100/PK으로도 사용됨
36 x 24 in. (91 x 61 cm)Translucent
카탈로그 번호 22032926
FISHERMIS-22032926-100/PK으로도 사용됨
제품 가격(KRW)
-
치수(길이 x 폭):
36 x 24 in. (91 x 61 cm)
색상:
Translucent
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners
사양
용량(미터법)170 L
인증/적합성ASTM D1709-98
마개 유형Zip Tie
디자인Non-printed without Sterilization Patch
치수(길이 x 폭)36 x 24 in. (91 x 61 cm)
일회용Single-use
용도(애플리케이션)Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
길이(영국식 단위)36 in.
길이(미터법)91 cm
멸균Non-sterile
두께1.5 mil (0.03 mm)
폭(영국식 단위)24 in.
폭(미터법)61 cm
오토클레이브 가능Autoclavable
용량(영국식 단위)20 gal.
색상Translucent
물질Polypropylene
제품 유형Biohazard Bag
Unit SizePack of 100