Citric acid, 99.5%, for analysis, anhydrous
Citric acid, 99.5%, for analysis, anhydrous
Citric acid, 99.5%, for analysis, anhydrous
Thermo Scientific Chemicals

Citric acid, 99.5%, for analysis, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 220341000
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.60
Save 10.95 (14%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.60
Save 10.95 (14%)
Each
Chemical Identifiers
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Chloride (Cl)=<10 ppm
Sulfate (SO4)=<100 ppm
Iron (Fe)=<5 ppm
Substance darkened by H2SO4Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
RUO – Research Use Only