Ethylenediamine, 99+%
Ethylenediamine, 99+%
Ethylenediamine, 99+%
Thermo Scientific Chemicals

Ethylenediamine, 99+%

CAS: 107-15-3 | C2H8N2 | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 220420010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-15-3
IUPAC Nameethane-1,2-diamine
Molecular FormulaC2H8N2
InChI KeyPIICEJLVQHRZGT-UHFFFAOYSA-N
SMILESNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (Coulometric)
Refractive index1.4555 to 1.4575 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only