Methyl salicylate, 99%
Methyl salicylate, 99%
Methyl salicylate, 99%
Thermo Scientific Chemicals

Methyl salicylate, 99%

CAS: 119-36-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 220490025
Price (USD)
224.65
Online exclusive
249.00
Save 24.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
224.65
Online exclusive
249.00
Save 24.35
Each
Chemical Identifiers
CAS119-36-8
IUPAC Namemethyl 2-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyOSWPMRLSEDHDFF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5350 to 1.5380 (20°C, 589 nm)
Density(20/4°C) 1.180 to 1.186 g/ml
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only