Cell MAb Medium, animal component free, is a complete medium that contains L-glutamine and is supplemented with 0.3% Difco SelectRead more
Have Questions?
Catalog Number
Quantity
220513
1000 mL
Catalog number 220513
Price (USD)
107.00
Each
Quantity:
1000 mL
Price (USD)
107.00
Each
Cell MAb Medium, animal component free, is a complete medium that contains L-glutamine and is supplemented with 0.3% Difco Select Soytone, an enzymatic digest of soy. This formulation does not contain phenol red, Pluronic® F68 (Kolliphor® P188), or other surfactants. It does not contain attachment factors, which may need to be added if this medium is used with adherent cell lines.
Specifications
Product TypeMAb Medium
Quantity1000 mL
With AdditivesGlutamine
Without AdditivesNo Phenol Red, No Pluronic Acid, No Other Surfactants
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C. Protect from light.